Alfa Aesar™ Methyl 3-amino-4-methylthiophene-2-carboxylate, 98%

Manufacturer: Alfa Aesar™ A1011514

Quantity 25g

 View more versions of this product

Catalog No. AAA1011514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85006-31-1
    Molecular Formula C7H9NO2S
    Formula Weight 171.22
    MDL Number MFCD00051822
    Beilstein 973196
    Synonym 3-Amino-4-methylthiophene-2-carboxylic acid methyl ester
    Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate

    Specifications

    Formula Weight 171.22
    Molecular Formula C7H9NO2S
    CAS 85006-31-1
    Quantity 25g
    MDL Number MFCD00051822
    Beilstein 973196
    Synonym 3-Amino-4-methylthiophene-2-carboxylic acid methyl ester
    Melting Point 84° to 88°C
    Flash Point >100°C (212°F)
    Chemical Name or Material Methyl 3-amino-4-methylthiophene-2-carboxylate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit