Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-aminocrotonate, 97%

Click to view available options
Quantity:
100g
500g
14205-39-1
C5H9NO2
115.132
MFCD00008072
XKORCTIIRYKLLG-ARJAWSKDSA-N
methyl 3-aminocrotonate,methyl 3-aminobut-2-enoate,methyl 2z-3-aminobut-2-enoate,z-methyl 3-aminobut-2-enoate,3-aminocrotonic acid methyl ester,methyl beta-aminocrotonate,methyl z-3-aminobut-2-enoate,methyl 2z-3-amino-2-butenoate,beta-aminocrotonic acid methyl ester,3-amino-2-butenoic acid methyl ester
643918
methyl (Z)-3-aminobut-2-enoate
CC(=CC(=O)OC)N
14205-39-1
C5H9NO2
115.132
MFCD00008072
XKORCTIIRYKLLG-ARJAWSKDSA-N
methyl 3-aminocrotonate,methyl 3-aminobut-2-enoate,methyl 2z-3-aminobut-2-enoate,z-methyl 3-aminobut-2-enoate,3-aminocrotonic acid methyl ester,methyl beta-aminocrotonate,methyl z-3-aminobut-2-enoate,methyl 2z-3-amino-2-butenoate,beta-aminocrotonic acid methyl ester,3-amino-2-butenoic acid methyl ester
643918
methyl (Z)-3-aminobut-2-enoate
CC(=CC(=O)OC)N
Boiling Point 122°C (42mmHg)
Quantity 500g
Melting Point 81°C to 85°C
Flash Point 91°C (195°F)
Beilstein 956592
Formula Weight 115.13
Percent Purity 97%
Chemical Name or Material Methyl 3-aminocrotonate

GHS H Statement
H302-H332-H315-H319-H317-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EM9092500

TSCA : TSCA