Alfa Aesar™ Methyl 3-aminocrotonate, 97%

Manufacturer: Alfa Aesar™ B2495322

Quantity 100g

 View more versions of this product

Catalog No. AAB2495322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14205-39-1
    Molecular Formula C5H9NO2
    Formula Weight 115.13
    MDL Number MFCD00008072
    Beilstein 956592
    Synonym 3-Aminocrotonic acid methyl ester; Methyl 3-aminobut-2-enoate
    Chemical Name or Material Methyl 3-aminocrotonate

    Specifications

    Formula Weight 115.13
    TSCA TSCA
    MDL Number MFCD00008072
    Health Hazard 1 Warning
    Melting Point 81° to 85°C
    Boiling Point 122°C (42mmHg)
    Quantity 100g
    Flash Point 91°C (195°F)
    Health Hazard 2 GHS H Statement
    H302-H332-H315-H319-H317-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number EM9092500
    Molecular Formula C5H9NO2
    CAS 14205-39-1
    Beilstein 956592
    Synonym 3-Aminocrotonic acid methyl ester; Methyl 3-aminobut-2-enoate
    Chemical Name or Material Methyl 3-aminocrotonate
    Assay 97%

    GHS H Statement
    H302-H332-H315-H319-H317-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit