Alfa Aesar™ Methyl 3-bromo-2-fluorobenzoate, 98%

Manufacturer: Alfa Aesar™ H3686506

Quantity 5g

 View more versions of this product

Catalog No. AAH3686506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 206551-41-9
    Formula Weight 233.04
    MDL Number MFCD09261255
    Synonym 3-Bromo-2-fluorobenzoic acid methyl ester
    Chemical Name or Material Methyl 3-bromo-2-fluorobenzoate

    Specifications

    Formula Weight 233.04
    Melting Point 35° to 37°C
    Physical Form Fused Solid
    MDL Number MFCD09261255
    Synonym 3-Bromo-2-fluorobenzoic acid methyl ester
    Color White to cream
    CAS 206551-41-9
    Quantity 5g
    Chemical Name or Material Methyl 3-bromo-2-fluorobenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit