Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-bromo-4-methylbenzoate, 99%

Click to view available options
Quantity:
5g
25g
100g
104901-43-1
C9H9BrO2
229.073
MFCD00144769
MASRAGFWFYHMFI-UHFFFAOYSA-N
3-bromo-4-methylbenzoic acid methyl ester,methyl 3-bromo-p-toluate,methyl3-bromo-4-methylbenzoate,benzoic acid, 3-bromo-4-methyl-, methyl ester,3-bromo-p-toluic acid methyl ester,3-bromo-4-methyl methyl benzoate,3-bromo-4-methyl-benzoic acid methyl ester,pubchem3967,acmc-2098dw,methyl 3-bromo4-methylbenzoate
7015747
methyl 3-bromo-4-methylbenzoate
CC1=C(C=C(C=C1)C(=O)OC)Br
104901-43-1
C9H9BrO2
229.073
MFCD00144769
MASRAGFWFYHMFI-UHFFFAOYSA-N
3-bromo-4-methylbenzoic acid methyl ester,methyl 3-bromo-p-toluate,methyl3-bromo-4-methylbenzoate,benzoic acid, 3-bromo-4-methyl-, methyl ester,3-bromo-p-toluic acid methyl ester,3-bromo-4-methyl methyl benzoate,3-bromo-4-methyl-benzoic acid methyl ester,pubchem3967,acmc-2098dw,methyl 3-bromo4-methylbenzoate
7015747
methyl 3-bromo-4-methylbenzoate
CC1=C(C=C(C=C1)C(=O)OC)Br
Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Quantity 25g
Formula Weight 229.07
Percent Purity 99%
Chemical Name or Material Methyl 3-bromo-4-methylbenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning