Alfa Aesar™ Methyl 3-bromo-5-hydroxybenzoate, 97%

Manufacturer: Alfa Aesar™ H51724MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51724MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 192810-12-1
    Molecular Formula C8H7BrO3
    Formula Weight 231.05
    MDL Number MFCD10566800
    Synonym 3-Bromo-5-hydroxybenzoic acid methyl ester
    Chemical Name or Material Methyl 3-bromo-5-hydroxybenzoate

    Specifications

    Formula Weight 231.05
    Molecular Formula C8H7BrO3
    MDL Number MFCD10566800
    Synonym 3-Bromo-5-hydroxybenzoic acid methyl ester
    Health Hazard 1 Warning
    Melting Point 130° to 135°C
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    CAS 192810-12-1
    Chemical Name or Material Methyl 3-bromo-5-hydroxybenzoate
    Assay 97%

    Description

    Applications
    It is employed as intermediate for pharmaceutical.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit