Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-bromobenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
618-89-3
C8H7BrO2
215.05
MFCD00017777
KMFJVYMFCAIRAN-UHFFFAOYSA-N
methyl m-bromobenzoate,3-bromobenzoic acid methyl ester,benzoic acid, 3-bromo-, methyl ester,3-bromobenzoic acid, methyl ester,methyl-3-bromobenzoate,benzoic acid, m-bromo-, methyl ester,m-bromobenzoic acid methyl ester,pubchem3961,methyl m-bromo benzoate,methyl 3-bromo-benzoate
12070
methyl 3-bromobenzoate
COC(=O)C1=CC(=CC=C1)Br
618-89-3
C8H7BrO2
215.05
MFCD00017777
KMFJVYMFCAIRAN-UHFFFAOYSA-N
methyl m-bromobenzoate,3-bromobenzoic acid methyl ester,benzoic acid, 3-bromo-, methyl ester,3-bromobenzoic acid, methyl ester,methyl-3-bromobenzoate,benzoic acid, m-bromo-, methyl ester,m-bromobenzoic acid methyl ester,pubchem3961,methyl m-bromo benzoate,methyl 3-bromo-benzoate
12070
methyl 3-bromobenzoate
COC(=O)C1=CC(=CC=C1)Br
Quantity 25g
Color White to Yellow
Packaging Glass bottle
Boiling Point 139°C to 140°C (35.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 215.05
Physical Form Crystalline Solid or Liquid
Percent Purity 98%
Chemical Name or Material Methyl 3-bromobenzoate

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-569-9

TSCA : TSCA