Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-chloro-4-methoxybenzoate 98%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
37908-98-8
C9H9ClO3
200.618
MFCD00017188
PINQDVFQCCFACD-UHFFFAOYSA-N
methyl 3-chloro-p-anisate,3-chloro-4-methoxybenzoic acid methyl ester,methyl3-chloro-4-methoxybenzoate,benzoic acid, 3-chloro-4-methoxy-, methyl ester,methyl-3-chlor-anisat,acmc-1aehy,rarechem al bf 0700,methyl-3-chloro-4-methoxybenzoate,2-chloro-4-methoxycarbonyl anisole,3-cl-4-ch3o-c6h3-cooch3
123469
methyl 3-chloro-4-methoxybenzoate
COC1=C(C=C(C=C1)C(=O)OC)Cl
37908-98-8
C9H9ClO3
200.618
MFCD00017188
PINQDVFQCCFACD-UHFFFAOYSA-N
methyl 3-chloro-p-anisate,3-chloro-4-methoxybenzoic acid methyl ester,methyl3-chloro-4-methoxybenzoate,benzoic acid, 3-chloro-4-methoxy-, methyl ester,methyl-3-chlor-anisat,acmc-1aehy,rarechem al bf 0700,methyl-3-chloro-4-methoxybenzoate,2-chloro-4-methoxycarbonyl anisole,3-cl-4-ch3o-c6h3-cooch3
123469
methyl 3-chloro-4-methoxybenzoate
COC1=C(C=C(C=C1)C(=O)OC)Cl
Packaging Glass bottle
Quantity 50g
Melting Point 93°C to 97°C
Infrared Spectrum Authentic
Color White to Yellow
Assay Percent Range 98%
Linear Formula CH3OC6H3(Cl)COOCH3
Formula Weight 200.62
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Methyl 3-chloro-4-methoxybenzoate
Show More Show Less