Learn More
Alfa Aesar™ Methyl 3-cyano-5-fluorobenzoate, 98%
Manufacturer:  Alfa Aesar™ H31759MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH31759MD
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Chemical Identifiers
886732-29-2
179.15
3-Cyano-5-fluorobenzoic acid methyl ester
C9H6FNO2
MFCD08688582
Methyl 3-cyano-5-fluorobenzoate
Specifications
886732-29-2
179.15
3-Cyano-5-fluorobenzoic acid methyl ester
Methyl 3-cyano-5-fluorobenzoate
C9H6FNO2
MFCD08688582
250mg
98%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning