Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-cyclopentenecarboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
58101-60-3
C7H10O2
126.16
CEOILRYKIJRPBZ-UHFFFAOYSA-N
methyl 3-cyclopentenecarboxylate,methyl cyclopent-3-enecarboxylate,methyl-3-cyclopentene-1-carboxylate,methyl3-cyclopentenecarboxylate,3-cyclopentene-1-carboxylic acid methyl ester,3-cyclopentene-1-carboxylic acid, methyl ester,acmc-209m2j,methyl cyclopent-3-ene carboxylate,4-methoxycarbonyl cyclopent-1-ene,methyl 3-cyclopentene-1-carboxylate
1514118
methyl cyclopent-3-ene-1-carboxylate
COC(=O)C1CC=CC1
58101-60-3
C7H10O2
126.16
CEOILRYKIJRPBZ-UHFFFAOYSA-N
methyl 3-cyclopentenecarboxylate,methyl cyclopent-3-enecarboxylate,methyl-3-cyclopentene-1-carboxylate,methyl3-cyclopentenecarboxylate,3-cyclopentene-1-carboxylic acid methyl ester,3-cyclopentene-1-carboxylic acid, methyl ester,acmc-209m2j,methyl cyclopent-3-ene carboxylate,4-methoxycarbonyl cyclopent-1-ene,methyl 3-cyclopentene-1-carboxylate
1514118
methyl cyclopent-3-ene-1-carboxylate
COC(=O)C1CC=CC1
Packaging Glass Bottle
Quantity 1g
Flash Point 49°C
Infrared Spectrum Authentic
Boiling Point 149.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material Methyl 3-cyclopentenecarboxylate

GHS H Statement
Flammable liquid and vapour.
Causes serious eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger