Alfa Aesar™ Methyl 3-fluoro-2-methoxybenzoate, 98%

Manufacturer: Alfa Aesar™ H3214703

Quantity 1g

 View more versions of this product

Catalog No. AAH3214703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106428-04-0
    Molecular Formula C9H9FO3
    Formula Weight 184.17
    MDL Number MFCD09753133
    Synonym 3-Fluoro-2-methoxybenzoic acid methyl ester
    Chemical Name or Material Methyl 3-fluoro-2-methoxybenzoate

    Specifications

    Formula Weight 184.17
    Molecular Formula C9H9FO3
    MDL Number MFCD09753133
    CAS 106428-04-0
    Synonym 3-Fluoro-2-methoxybenzoic acid methyl ester
    Quantity 1g
    Chemical Name or Material Methyl 3-fluoro-2-methoxybenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit