Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-fluorophenylacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
64123-77-9
C9H9FO2
168.17
MFCD03093288
OXYLCGRXCQARQV-UHFFFAOYSA-N
methyl 2-3-fluorophenyl acetate,methyl 3-fluorophenylacetate,methyl3-fluorophenylacetate,methyl 3-fluorophenyl acetate,3-fluorobenzeneacetic acid methyl ester,benzeneacetic acid, 3-fluoro-, methyl ester,3-fluorophenylacetic acid methyl ester,3-fluorophenyl acetic acid methyl ester,3-fluoro-phenyl-acetic acid methyl ester,2-3-fluorophenyl acetic acid methyl ester
2734108
methyl 2-(3-fluorophenyl)acetate
COC(=O)CC1=CC(=CC=C1)F
64123-77-9
C9H9FO2
168.17
MFCD03093288
OXYLCGRXCQARQV-UHFFFAOYSA-N
methyl 2-3-fluorophenyl acetate,methyl 3-fluorophenylacetate,methyl3-fluorophenylacetate,methyl 3-fluorophenyl acetate,3-fluorobenzeneacetic acid methyl ester,benzeneacetic acid, 3-fluoro-, methyl ester,3-fluorophenylacetic acid methyl ester,3-fluorophenyl acetic acid methyl ester,3-fluoro-phenyl-acetic acid methyl ester,2-3-fluorophenyl acetic acid methyl ester
2734108
methyl 2-(3-fluorophenyl)acetate
COC(=O)CC1=CC(=CC=C1)F
Refractive Index 1.4870 to 1.489
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H4CH2CO2H
Formula Weight 168.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl 3-fluorophenylacetate, 98%
Show More Show Less