Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-(furfurylthio)propionate, 98%

Manufacturer:  Alfa Aesar™ H3241003

Quantity 1g

 View more versions of this product

Catalog No. AAH3241003


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

94278-26-9
200.26
3-(Furfurylthio)propionic acid methyl ester
C9H12O3S
MFCD00053109
Methyl 3-(furfurylthio)propionate

Specifications

94278-26-9
200.26
3-(Furfurylthio)propionic acid methyl ester
1g
Methyl 3-(furfurylthio)propionate
C9H12O3S
MFCD00053109
114° to 116°C (1.12mmHg)
92°C (198°F)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit