Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-hydroxy-4-iodobenzoate, 97%

Click to view available options
Quantity:
250mg
1g
5g
157942-12-6
C8H7IO3
278.045
MFCD06203960
LXCQVWRESZDFGW-UHFFFAOYSA-N
methyl-4-iodo-3-hydroxy benzoate,3-hydroxy-4-iodobenzoic acid methyl ester,3-hydroxy-4-iodo-benzoic acid methyl ester,methyl 3-hydroxy-4-iodobenzenecarboxylate,benzoic acid, 3-hydroxy-4-iodo-, methyl ester,rarechem al bf 0964,methyl 3-hydroxy4-iodobenzoate,methyl 4-iodo-3-hydroxybenzoate,methyl-3-hydroxy-4-iodobenzoate,methyl-4-iodo-3-hydroxybenzoate
10378812
methyl 3-hydroxy-4-iodobenzoate
COC(=O)C1=CC(=C(C=C1)I)O
157942-12-6
C8H7IO3
278.045
MFCD06203960
LXCQVWRESZDFGW-UHFFFAOYSA-N
methyl-4-iodo-3-hydroxy benzoate,3-hydroxy-4-iodobenzoic acid methyl ester,3-hydroxy-4-iodo-benzoic acid methyl ester,methyl 3-hydroxy-4-iodobenzenecarboxylate,benzoic acid, 3-hydroxy-4-iodo-, methyl ester,rarechem al bf 0964,methyl 3-hydroxy4-iodobenzoate,methyl 4-iodo-3-hydroxybenzoate,methyl-3-hydroxy-4-iodobenzoate,methyl-4-iodo-3-hydroxybenzoate
10378812
methyl 3-hydroxy-4-iodobenzoate
COC(=O)C1=CC(=C(C=C1)I)O
Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Quantity 1g
Sensitivity Light Sensitive
Beilstein 7289690
Formula Weight 278.05
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxy-4-iodobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning