Alfa Aesar™ Methyl 3-hydroxythiophene-2-carboxylate, 97%

Manufacturer: Alfa Aesar™ B2212914

Quantity 25g

 View more versions of this product

Catalog No. AAB2212914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5118-06-9
    Molecular Formula C6H6O3S
    Formula Weight 158.18
    MDL Number MFCD00055642
    Synonym 3-Hydroxythiophene-2-carboxylic acid methyl ester
    Chemical Name or Material Methyl 3-hydroxythiophene-2-carboxylate

    Specifications

    Formula Weight 158.18
    Molecular Formula C6H6O3S
    Boiling Point 107° to 109°C (13mmHg)
    Quantity 25g
    MDL Number MFCD00055642
    CAS 5118-06-9
    Synonym 3-Hydroxythiophene-2-carboxylic acid methyl ester
    Melting Point 44° to 48°C
    Chemical Name or Material Methyl 3-hydroxythiophene-2-carboxylate
    Assay 97%

    Description

    Applications
    Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit