Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-hydroxythiophene-2-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
25g
5144-10-5
C12H15N
173.26
MFCD00055642
NCDPYDGGUAKABA-UHFFFAOYSA-N
pentamethylbenzonitrile,6-aminobutyric acid,pentamethylbenzonitril,acmc-1axcs,benzonitrile,2,3,4,5,6-pentamethyl,2,3,4,5,6-pentamethylbenzenecarbonitrile
78824
2,3,4,5,6-pentamethylbenzonitrile
CC1=C(C(=C(C(=C1C)C)C#N)C)C
5144-10-5
C12H15N
173.26
MFCD00055642
NCDPYDGGUAKABA-UHFFFAOYSA-N
pentamethylbenzonitrile,6-aminobutyric acid,pentamethylbenzonitril,acmc-1axcs,benzonitrile,2,3,4,5,6-pentamethyl,2,3,4,5,6-pentamethylbenzenecarbonitrile
78824
2,3,4,5,6-pentamethylbenzonitrile
CC1=C(C(=C(C(=C1C)C)C#N)C)C
Applications
Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Melting Point 44°C to 48°C
Boiling Point 107°C to 109°C (13mmHg)
Quantity 5g
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxythiophene-2-carboxylate