Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-iodo-4-methoxybenzoate, 98%

Manufacturer:  Alfa Aesar™ A1054922

Quantity 100g

 View more versions of this product

Catalog No. AAA1054922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

35387-93-0
292.07
2692711
Methyl 3-iodo-4-methoxybenzoate
C9H9IO3
MFCD00052925
3-Iodo-4-methoxybenzoic acid methyl ester

Specifications

35387-93-0
292.07
2692711
93° to 95°C
Methyl 3-iodo-4-methoxybenzoate
C9H9IO3
MFCD00052925
3-Iodo-4-methoxybenzoic acid methyl ester
100g
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit