Alfa Aesar™ Methyl 3-methoxy-4-methylbenzoate, 98%

Manufacturer: Alfa Aesar™ B2269503

Quantity 1g

 View more versions of this product

Catalog No. AAB2269503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3556-83-0
    Molecular Formula C10H12O3
    Formula Weight 180.21
    MDL Number MFCD00082710
    Beilstein 1948339
    Synonym 3-Methoxy-4-methylbenzoic acid methyl ester; Methyl 3-methoxy-p-toluate
    Chemical Name or Material Methyl 3-methoxy-4-methylbenzoate

    Specifications

    Formula Weight 180.21
    Quantity 1g
    Molecular Formula C10H12O3
    MDL Number MFCD00082710
    CAS 3556-83-0
    Beilstein 1948339
    Synonym 3-Methoxy-4-methylbenzoic acid methyl ester; Methyl 3-methoxy-p-toluate
    Melting Point 47° to 50°C
    Boiling Point 119° to 120°C (1mmHg)
    Flash Point >110°C (230°F)
    Chemical Name or Material Methyl 3-methoxy-4-methylbenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit