Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-methoxy-4-methylbenzoate, 98%

Click to view available options
Quantity:
1g
5g
25g
3556-83-0
C10H12O3
180.203
MFCD00082710
LLEXCSBUSVRBCA-UHFFFAOYSA-N
3-methoxy-4-methylbenzoic acid methyl ester,methyl 3-methoxy-p-toluate,4-methyl-m-anisic acid methyl ester,benzoic acid, 3-methoxy-4-methyl-, methyl ester,pubchem13650,methyl 4-methyl-m-anisate,acmc-1cs22,methyl3-methoxy-4-methylbenzoate,methyl 3-methoxy-4-methyl-benzoate,3-methoxy-p-toluic acid methyl ester
591123
methyl 3-methoxy-4-methylbenzoate
CC1=C(C=C(C=C1)C(=O)OC)OC
3556-83-0
C10H12O3
180.203
MFCD00082710
LLEXCSBUSVRBCA-UHFFFAOYSA-N
3-methoxy-4-methylbenzoic acid methyl ester,methyl 3-methoxy-p-toluate,4-methyl-m-anisic acid methyl ester,benzoic acid, 3-methoxy-4-methyl-, methyl ester,pubchem13650,methyl 4-methyl-m-anisate,acmc-1cs22,methyl3-methoxy-4-methylbenzoate,methyl 3-methoxy-4-methyl-benzoate,3-methoxy-p-toluic acid methyl ester
591123
methyl 3-methoxy-4-methylbenzoate
CC1=C(C=C(C=C1)C(=O)OC)OC
Flash Point >110°C (230°F)
Boiling Point 119°C to 120°C (1mmHg)
Quantity 25g
Melting Point 47°C to 50°C
Beilstein 1948339
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material Methyl 3-methoxy-4-methylbenzoate