Alfa Aesar™ Methyl 3-methoxyacrylate, 95+%

Manufacturer: Alfa Aesar™ B2295014

Quantity 25g

 View more versions of this product

Catalog No. AAB2295014

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 34846-90-7
    Molecular Formula CH3OCH≡CHCO2CH3
    Formula Weight 116.13
    MDL Number MFCD00128206
    Synonym 3-Methoxyacrylic acid methyl ester; Methyl 3-methoxypropenoate
    Chemical Name or Material Methyl 3-methoxyacrylate

    Specifications

    Formula Weight 116.13
    Molecular Formula CH3OCH≡CHCO2CH3
    MDL Number MFCD00128206
    Boiling Point 168° to 170°C
    Quantity 25g
    Flash Point 65°C (149°F)
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P363-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 34846-90-7
    Synonym 3-Methoxyacrylic acid methyl ester; Methyl 3-methoxypropenoate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317-H227
    May cause an allergic skin reaction.
    Combustible liquid.
    Recommended Storage 2° to 8°C
    Chemical Name or Material Methyl 3-methoxyacrylate

    GHS H Statement
    H317-H227
    May cause an allergic skin reaction.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P261-P280-P363-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit