Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-methyl-2-furoate, 98%

Manufacturer:  Alfa Aesar™ A1054122

Quantity 100g

 View more versions of this product

Catalog No. AAA1054122


  Check Availability
Add to cart

Specifications

Description

Applications
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

Solubility
Insoluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

6141-57-7
140.14
116835
Methyl 3-methyl-2-furoate
C7H8O3
MFCD00014110
3-Methylfuran-2-carboxylic acid methyl ester; 3-Methyl-2-furoic acid methyl ester

Specifications

6141-57-7
140.14
116835
33° to 37°C
100g
Methyl 3-methyl-2-furoate
C7H8O3
MFCD00014110
3-Methylfuran-2-carboxylic acid methyl ester; 3-Methyl-2-furoic acid methyl ester
60°C (1mmHg)
82°C (180°F)
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit