Alfa Aesar™ Methyl 3-methylbenzo[b]thiophene-2-carboxylate, 97%

Manufacturer: Alfa Aesar™ B2406914

Quantity 25g

 View more versions of this product

Catalog No. AAB2406914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3133-81-1
    Molecular Formula C11H10O2S
    Formula Weight 206.26
    MDL Number MFCD02683110
    Synonym 3-Methylbenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-methylthianaphthene-2-carboxylate
    Chemical Name or Material Methyl 3-methylbenzo[b]thiophene-2-carboxylate

    Specifications

    Formula Weight 206.26
    Molecular Formula C11H10O2S
    MDL Number MFCD02683110
    Synonym 3-Methylbenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-methylthianaphthene-2-carboxylate
    Melting Point 102° to 105°C
    Quantity 25g
    CAS 3133-81-1
    Chemical Name or Material Methyl 3-methylbenzo[b]thiophene-2-carboxylate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit