Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-methylbenzo[b]thiophene-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ B2406903

Quantity 1g

 View more versions of this product

Catalog No. AAB2406903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3133-81-1
206.26
3-Methylbenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-methylthianaphthene-2-carboxylate
C11H10O2S
MFCD02683110
Methyl 3-methylbenzo[b]thiophene-2-carboxylate

Specifications

3133-81-1
206.26
3-Methylbenzo[b]thiophene-2-carboxylic acid methyl ester; Methyl 3-methylthianaphthene-2-carboxylate
1g
97%
C11H10O2S
MFCD02683110
102° to 105°C
Methyl 3-methylbenzo[b]thiophene-2-carboxylate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit