Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-methylphenylacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
53088-69-0
C10H12O2
164.2
MFCD00968468
AWTKFTNNPQGGLX-UHFFFAOYSA-N
methyl 2-m-tolyl acetate,methyl 3-methylphenylacetate,methyl 2-3-methylphenyl acetate,3-methylphenylacetic acid methyl ester,benzeneacetic acid, 3-methyl-, methyl ester,methyl 2-m-tolylacetate,m-tolylacetic acid methyl ester,m-toly-acetic acid methyl ester,m-tolyl-acetic acid methyl ester,methyl 2-3-methylphenyl ethanoate
5135288
methyl 2-(3-methylphenyl)acetate
CC1=CC(=CC=C1)CC(=O)OC
53088-69-0
C10H12O2
164.2
MFCD00968468
AWTKFTNNPQGGLX-UHFFFAOYSA-N
methyl 2-m-tolyl acetate,methyl 3-methylphenylacetate,methyl 2-3-methylphenyl acetate,3-methylphenylacetic acid methyl ester,benzeneacetic acid, 3-methyl-, methyl ester,methyl 2-m-tolylacetate,m-tolylacetic acid methyl ester,m-toly-acetic acid methyl ester,m-tolyl-acetic acid methyl ester,methyl 2-3-methylphenyl ethanoate
5135288
methyl 2-(3-methylphenyl)acetate
CC1=CC(=CC=C1)CC(=O)OC
Boiling Point 117°C (13.0mmHg)
Refractive Index 1.5045 to 1.5065
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3C6H4CH2COOCH3
Formula Weight 164.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl 3-methylphenylacetate
Show More Show Less