Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(methylthio)propionate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
13532-18-8
C5H10O2S
134.19
MFCD00008565
DMMJVMYCBULSIS-UHFFFAOYSA-N
methyl 3-methylthio propionate,methyl 3-methylthiopropionate,methyl 3-methylthio propanoate,propanoic acid, 3-methylthio-, methyl ester,methyl beta-methiopropionate,methyl 3-methylsulfanyl propanoate,methyl-3-methylthio propionate,methyl 3-methylthiopropanoate,propionic acid, 3-methylthio-, methyl ester,methyl beta-methylthiopropionate
61641
methyl 3-methylsulfanylpropanoate
COC(=O)CCSC
13532-18-8
C5H10O2S
134.19
MFCD00008565
DMMJVMYCBULSIS-UHFFFAOYSA-N
methyl 3-methylthio propionate,methyl 3-methylthiopropionate,methyl 3-methylthio propanoate,propanoic acid, 3-methylthio-, methyl ester,methyl beta-methiopropionate,methyl 3-methylsulfanyl propanoate,methyl-3-methylthio propionate,methyl 3-methylthiopropanoate,propionic acid, 3-methylthio-, methyl ester,methyl beta-methylthiopropionate
61641
methyl 3-methylsulfanylpropanoate
COC(=O)CCSC
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.0700g/mL
Specific Gravity 1.07
Boiling Point 74.0°C to 75.0°C (13.0mmHg)
Flash Point 72°C
Quantity 5g
Color Colorless
Refractive Index 1.4640 to 1.4660
Assay Percent Range 99+%
Linear Formula CH3SCH2CH2CO2CH3
Solubility Information Solubility in water: insoluble.
Formula Weight 134.19
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Methyl 3-(methylthio)propionate, 99+%
Show More Show Less

STENCH

EINECSNumber : 236-883-6

TSCA : TSCA