Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-nitrosalicylate, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
22621-41-6
C8H7NO5
197.15
MFCD00272263
NIBVYEHAFBEVFI-UHFFFAOYSA-N
methyl 2-hydroxy-3-nitrobenzoate,methyl 3-nitrosalicylate,benzoic acid, 2-hydroxy-3-nitro-, methyl ester,2-hydroxy-3-nitrobenzoic acid methyl ester,2-hydroxy-3-nitro-benzoic acid methyl ester,salicylic acid, 3-nitro-, methyl ester,methyl-3-nitrosalicylat,methyl-m-nitrosalicylate,acmc-209fxo,ksc490c4l
596904
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
22621-41-6
C8H7NO5
197.15
MFCD00272263
NIBVYEHAFBEVFI-UHFFFAOYSA-N
methyl 2-hydroxy-3-nitrobenzoate,methyl 3-nitrosalicylate,benzoic acid, 2-hydroxy-3-nitro-, methyl ester,2-hydroxy-3-nitrobenzoic acid methyl ester,2-hydroxy-3-nitro-benzoic acid methyl ester,salicylic acid, 3-nitro-, methyl ester,methyl-3-nitrosalicylat,methyl-m-nitrosalicylate,acmc-209fxo,ksc490c4l
596904
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])O
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Melting Point 127°C to 132°C
Quantity 5g
Assay Percent Range 97.5% min. (GC)
Linear Formula NO2C6H3(OH)COOCH3
Formula Weight 197.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Methyl 3-nitrosalicylate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning