Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
25g
188614-01-9
C10H9NO3S
223.246
MFCD00449104
UFSVRHNRNVVBJR-UHFFFAOYSA-N
methyl 3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate,methyl-3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate,methyl 3-oxo-3,4-dihydro-2h-benzo b 1,4 thiazine-6-carboxylate,methyl 3-oxo-2h-1,4-benzothiazine-6-carboxylate,methyl 3,4-dihydro-3-oxo-2h-benzo b 1,4 thiazine-6-carboxylate,methyl 3-oxo-2,4-dihydro-1,4-benzothiazine-6-carboxylate,pubchem13044,ksc497g3b,methyl-3-oxo-3,4-dihydro-2h-1,,methyl3-oxo-2h-1,4-benzothiazine-6-carboxylate
699501
methyl 3-oxo-4H-1,4-benzothiazine-6-carboxylate
COC(=O)C1=CC2=C(C=C1)SCC(=O)N2
188614-01-9
C10H9NO3S
223.246
MFCD00449104
UFSVRHNRNVVBJR-UHFFFAOYSA-N
methyl 3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate,methyl-3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate,methyl 3-oxo-3,4-dihydro-2h-benzo b 1,4 thiazine-6-carboxylate,methyl 3-oxo-2h-1,4-benzothiazine-6-carboxylate,methyl 3,4-dihydro-3-oxo-2h-benzo b 1,4 thiazine-6-carboxylate,methyl 3-oxo-2,4-dihydro-1,4-benzothiazine-6-carboxylate,pubchem13044,ksc497g3b,methyl-3-oxo-3,4-dihydro-2h-1,,methyl3-oxo-2h-1,4-benzothiazine-6-carboxylate
699501
methyl 3-oxo-4H-1,4-benzothiazine-6-carboxylate
COC(=O)C1=CC2=C(C=C1)SCC(=O)N2
Melting Point 97°C to 99°C
Quantity 25g
Beilstein 14311523
Formula Weight 223.25
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate

storageNote1 : -30° to -10°C