Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-thiopheneacetate, 98%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
58414-52-1
C7H8O2S
156.20
MFCD00052947
RZGRWVULDSXQSM-UHFFFAOYSA-N
methyl 3-thienylacetate, methyl thiophene-3-acetate, methyl 2-3-thienyl acetate, methyl 2-thiophen-3-yl acetate, 3-thiopheneacetic acid, methyl ester, methyl thiophen-3-acetate, methyl 3-thiopheneacetate, methylthiophene-3-acetate, acmc-20akew, methyl 3-thienylacetate #
94041
methyl 2-thiophen-3-ylacetate
COC(=O)CC1=CSC=C1
58414-52-1
C7H8O2S
156.20
MFCD00052947
RZGRWVULDSXQSM-UHFFFAOYSA-N
methyl 3-thienylacetate, methyl thiophene-3-acetate, methyl 2-3-thienyl acetate, methyl 2-thiophen-3-yl acetate, 3-thiopheneacetic acid, methyl ester, methyl thiophen-3-acetate, methyl 3-thiopheneacetate, methylthiophene-3-acetate, acmc-20akew, methyl 3-thienylacetate #
94041
methyl 2-thiophen-3-ylacetate
COC(=O)CC1=CSC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Boiling Point 125°C to 129°C (25mmHg)
Beilstein 4378596
Refractive Index 1.5215
Quantity 1g
Formula Weight 156.2
Percent Purity 98%
Chemical Name or Material Methyl 3-thiopheneacetate

EINECSNumber : 261-242-2

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only