Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(trifluoromethoxy)benzoate 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
148438-00-0
C9H7F3O3
220.147
MFCD00041005
YWSQRIKZJLRGPT-UHFFFAOYSA-N
methyl 3-trifluoromethoxy benzoate,benzoic acid,3-trifluoromethoxy-, methyl ester,acmc-1bxui,tpc-i176,ywsqrikzjlrgpt-uhfffaoysa,methyl3-trifluoromethoxy benzoate,methyl-3-trifluoromethoxy benzoate,3-trifluoromethoxy benzoic acid methyl ester,3-trifluoromethyloxy-benzoic acid methyl ester
519012
methyl 3-(trifluoromethoxy)benzoate
COC(=O)C1=CC(=CC=C1)OC(F)(F)F
148438-00-0
C9H7F3O3
220.147
MFCD00041005
YWSQRIKZJLRGPT-UHFFFAOYSA-N
methyl 3-trifluoromethoxy benzoate,benzoic acid,3-trifluoromethoxy-, methyl ester,acmc-1bxui,tpc-i176,ywsqrikzjlrgpt-uhfffaoysa,methyl3-trifluoromethoxy benzoate,methyl-3-trifluoromethoxy benzoate,3-trifluoromethoxy benzoic acid methyl ester,3-trifluoromethyloxy-benzoic acid methyl ester
519012
methyl 3-(trifluoromethoxy)benzoate
COC(=O)C1=CC(=CC=C1)OC(F)(F)F
Quantity 5g
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Assay Percent Range 98%
Linear Formula CF3OC6H4CO2CH3
Formula Weight 220.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Methyl 3-(trifluoromethoxy)benzoate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.