Alfa Aesar™ Methyl 3-(trifluoromethyl)phenylacetate, 95%

Manufacturer: Alfa Aesar™ H3288306

Quantity 5g

 View more versions of this product

Catalog No. AAH3288306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62451-84-7
    Molecular Formula C10H9F3O2
    Formula Weight 218.18
    MDL Number MFCD04039762
    Synonym 3-(Trifluoromethyl)phenylacetic acid methyl ester
    Chemical Name or Material Methyl 3-(trifluoromethyl)phenylacetate

    Specifications

    Formula Weight 218.18
    Molecular Formula C10H9F3O2
    MDL Number MFCD04039762
    CAS 62451-84-7
    Synonym 3-(Trifluoromethyl)phenylacetic acid methyl ester
    Boiling Point 211°C
    Quantity 5g
    Chemical Name or Material Methyl 3-(trifluoromethyl)phenylacetate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit