Alfa Aesar™ Methyl 4-(3-thienyl)benzoate, 97%

Manufacturer: Alfa Aesar™ H2738606

Quantity 5g

 View more versions of this product

Catalog No. AAH2738606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20608-91-7
    Molecular Formula C12H10O2S
    Formula Weight 218.27
    MDL Number MFCD05863312
    Synonym 4-(3-Thienyl)benzoic acid methyl ester; 4-(3-thienyl)benoic acid methyl ester
    Chemical Name or Material Methyl 4-(3-thienyl)benzoate

    Specifications

    Formula Weight 218.27
    Molecular Formula C12H10O2S
    MDL Number MFCD05863312
    Synonym 4-(3-Thienyl)benzoic acid methyl ester; 4-(3-thienyl)benoic acid methyl ester
    Melting Point 162° to 163°C
    Quantity 5g
    CAS 20608-91-7
    Chemical Name or Material Methyl 4-(3-thienyl)benzoate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit