Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4,5-dimethoxy-3-hydroxybenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
83011-43-2
C10H12O5
212.2
MFCD00598192
LCIFXEQPXQVBGL-UHFFFAOYSA-N
3,4-dimethoxy-5-hydroxybenzoic acid methyl ester,methyl 3,4-dimethoxy-5-hydroxybenzoate,methyl 4,5-dimethoxy-3-hydroxybenzoate,methyl 3,4-methoxy-5-hydroxybenzoate,methyl 5-hydroxy-3,4-dimethoxybenzoate,methyl 3,4-dimehtoxy-5-hydroxybenzoate,methyl 4?5-dimethoxy-3-hydroxybenzoate,3,4-dimethoxy-5-hydroxybenzoate,methyl 3,4-dimethoxy-5-hydroxy-benzoate,methyl 3-hydroxy-4,5-dimethoxy-benzoate
2733956
methyl 3-hydroxy-4,5-dimethoxybenzoate
COC1=CC(=CC(=C1OC)O)C(=O)OC
83011-43-2
C10H12O5
212.2
MFCD00598192
LCIFXEQPXQVBGL-UHFFFAOYSA-N
3,4-dimethoxy-5-hydroxybenzoic acid methyl ester,methyl 3,4-dimethoxy-5-hydroxybenzoate,methyl 4,5-dimethoxy-3-hydroxybenzoate,methyl 3,4-methoxy-5-hydroxybenzoate,methyl 5-hydroxy-3,4-dimethoxybenzoate,methyl 3,4-dimehtoxy-5-hydroxybenzoate,methyl 4?5-dimethoxy-3-hydroxybenzoate,3,4-dimethoxy-5-hydroxybenzoate,methyl 3,4-dimethoxy-5-hydroxy-benzoate,methyl 3-hydroxy-4,5-dimethoxy-benzoate
2733956
methyl 3-hydroxy-4,5-dimethoxybenzoate
COC1=CC(=CC(=C1OC)O)C(=O)OC
Quantity 5g
Melting Point 80°C to 84°C
Packaging Glass bottle
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 212.2
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material Methyl 4, 5-dimethoxy-3-hydroxybenzoate, 98%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning