Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-amino-3-bromobenzoate, 97%

Click to view available options
Quantity:
1g
5g
106896-49-5
C8H8BrNO2
230.061
MFCD01861385
AIUWAOALZYWQBX-UHFFFAOYSA-N
4-amino-3-bromo-benzoic acid methyl ester,methyl4-amino-3-bromobenzoate,2-bromo-4-methoxycarbonylaniline,benzoic acid, 4-amino-3-bromo-, methyl ester,4-amino-3-bromobenzoic acid methyl ester,acmc-2098mp,methyl-4-amino-3-bromobenzoate,methyl 4-amino-3-bromo-benzoate,timtec-bb sbb012847,2-bromo-4-methoxycarbonyl aniline
1515280
methyl 4-amino-3-bromobenzoate
COC(=O)C1=CC(=C(C=C1)N)Br
106896-49-5
C8H8BrNO2
230.061
MFCD01861385
AIUWAOALZYWQBX-UHFFFAOYSA-N
4-amino-3-bromo-benzoic acid methyl ester,methyl4-amino-3-bromobenzoate,2-bromo-4-methoxycarbonylaniline,benzoic acid, 4-amino-3-bromo-, methyl ester,4-amino-3-bromobenzoic acid methyl ester,acmc-2098mp,methyl-4-amino-3-bromobenzoate,methyl 4-amino-3-bromo-benzoate,timtec-bb sbb012847,2-bromo-4-methoxycarbonyl aniline
1515280
methyl 4-amino-3-bromobenzoate
COC(=O)C1=CC(=C(C=C1)N)Br
Quantity 5g
Melting Point 105°C to 109°C
Beilstein 4905039
Formula Weight 230.06
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Methyl 4-amino-3-bromobenzoate

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger