Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-aminonicotinate, 98%

Click to view available options
Quantity:
1g
5g
16135-36-7
C7H8N2O2
152.153
MFCD00955996
OCARFFAPQGYGBP-UHFFFAOYSA-N
methyl 4-aminonicotinate,4-aminonicotinic acid methyl ester,4-amino-nicotinic acid methyl ester,4-aminopyridine-3-carboxylic acid methyl ester,3-pyridinecarboxylic acid, 4-amino-, methyl ester,nicotinic acid, 4-amino-, methyl ester,4-aminonicotinicacidmethylester,methyl-4-aminonicotinate,ksc493a3l,tpc-py097
592353
methyl 4-aminopyridine-3-carboxylate
COC(=O)C1=C(C=CN=C1)N
16135-36-7
C7H8N2O2
152.153
MFCD00955996
OCARFFAPQGYGBP-UHFFFAOYSA-N
methyl 4-aminonicotinate,4-aminonicotinic acid methyl ester,4-amino-nicotinic acid methyl ester,4-aminopyridine-3-carboxylic acid methyl ester,3-pyridinecarboxylic acid, 4-amino-, methyl ester,nicotinic acid, 4-amino-, methyl ester,4-aminonicotinicacidmethylester,methyl-4-aminonicotinate,ksc493a3l,tpc-py097
592353
methyl 4-aminopyridine-3-carboxylate
COC(=O)C1=C(C=CN=C1)N
Melting Point 172°C to 174°C
Quantity 1g
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material Methyl 4-aminonicotinate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning