Alfa Aesar™ Methyl 4-bromo-2-(bromomethyl)benzoate, 96%

Manufacturer: Alfa Aesar™ H52350MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52350MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78471-43-9
    Molecular Formula C9H8Br2O2
    Formula Weight 307.97
    MDL Number MFCD04114319
    Synonym 4-Bromo-2-(bromomethyl)benzoic acid methyl ester
    Chemical Name or Material Methyl 4-bromo-2-(bromomethyl)benzoate

    Specifications

    Formula Weight 307.97
    Molecular Formula C9H8Br2O2
    MDL Number MFCD04114319
    CAS 78471-43-9
    Synonym 4-Bromo-2-(bromomethyl)benzoic acid methyl ester
    Melting Point 76° to 78°C
    Quantity 250mg
    Chemical Name or Material Methyl 4-bromo-2-(bromomethyl)benzoate
    Assay 96%

    Description

    Applications
    Methyl 4-bromo-2-(bromomethyl)benzoate is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit