Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-bromo-2-(bromomethyl)benzoate, 96%

Manufacturer:  Alfa Aesar™ H5235003

Quantity 1g

 View more versions of this product

Catalog No. AAH5235003


  Check Availability
Add to cart

Specifications

Description

Applications
Methyl 4-bromo-2-(bromomethyl)benzoate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

78471-43-9
307.97
4-Bromo-2-(bromomethyl)benzoic acid methyl ester
C9H8Br2O2
MFCD04114319
Methyl 4-bromo-2-(bromomethyl)benzoate

Specifications

78471-43-9
307.97
4-Bromo-2-(bromomethyl)benzoic acid methyl ester
1g
96%
C9H8Br2O2
MFCD04114319
76° to 78°C
Methyl 4-bromo-2-(bromomethyl)benzoate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit