Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-bromo-3-methylbenzoate, 97%

Click to view available options
Quantity:
1g
5g
25g
148547-19-7
C9H9BrO2
229.073
MFCD00673014
GTZTYNPAPQKIIR-UHFFFAOYSA-N
4-bromo-3-methylbenzoic acid methyl ester,methyl4-bromo-3-methylbenzoate,methyl 4-bromo-m-toluate,4-bromo-3-methyl-benzoic acid methyl ester,methyl 4-bromo-3-methyl-benzoate,4-bromo-m-toluic acid methyl ester,benzoic acid, 4-bromo-3-methyl-, methyl ester,2-bromo-5-methoxycarbonyl toluene,3-methyl-4-bromobenzoic acid methyl ester,pubchem3970
3294148
methyl 4-bromo-3-methylbenzoate
CC1=C(C=CC(=C1)C(=O)OC)Br
148547-19-7
C9H9BrO2
229.073
MFCD00673014
GTZTYNPAPQKIIR-UHFFFAOYSA-N
4-bromo-3-methylbenzoic acid methyl ester,methyl4-bromo-3-methylbenzoate,methyl 4-bromo-m-toluate,4-bromo-3-methyl-benzoic acid methyl ester,methyl 4-bromo-3-methyl-benzoate,4-bromo-m-toluic acid methyl ester,benzoic acid, 4-bromo-3-methyl-, methyl ester,2-bromo-5-methoxycarbonyl toluene,3-methyl-4-bromobenzoic acid methyl ester,pubchem3970
3294148
methyl 4-bromo-3-methylbenzoate
CC1=C(C=CC(=C1)C(=O)OC)Br
Quantity 1g
Melting Point 39°C to 44°C
Beilstein 6593534
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material Methyl 4-bromo-3-methylbenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures