Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4'-bromobiphenyl-4-carboxylate, 95%

Manufacturer:  Alfa Aesar™ H5192006

Quantity 5g

 View more versions of this product

Catalog No. AAH5192006

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
It is employed in the design, synthesis, and biological evaluation of novel nonsteroidal farnesoid x receptor (fxr) antagonists.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

89901-03-1
291.14
4'-Bromobiphenyl-4-carboxylic acid methyl ester
C14H11BrO2
MFCD00496638
Methyl 4'-bromobiphenyl-4-carboxylate

Specifications

89901-03-1
291.14
4'-Bromobiphenyl-4-carboxylic acid methyl ester
Methyl 4'-bromobiphenyl-4-carboxylate
C14H11BrO2
MFCD00496638
5g
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit