Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-bromopyridine-2-carboxylate, ≥97%

Click to view available options
Quantity:
1g
5g
25g
29681-42-3
C7H6BrNO2
216.034
MFCD07374897
JZFLATQBIPILFS-UHFFFAOYSA-N
methyl 4-bromopicolinate,4-bromo-pyridine-2-carboxylic acid methyl ester,methyl 4-bromo-2-pyridinecarboxylate,methyl4-bromopicolinate,2-pyridinecarboxylic acid, 4-bromo-, methyl ester,4-bromopyridine-2-carboxylic acid methyl ester,abbypharma ap-10-5912,pubchem12733,acmc-209hag,methyl 4-bromo-pyridine-2-carboxylate
12828947
methyl 4-bromopyridine-2-carboxylate
COC(=O)C1=NC=CC(=C1)Br
29681-42-3
C7H6BrNO2
216.034
MFCD07374897
JZFLATQBIPILFS-UHFFFAOYSA-N
methyl 4-bromopicolinate,4-bromo-pyridine-2-carboxylic acid methyl ester,methyl 4-bromo-2-pyridinecarboxylate,methyl4-bromopicolinate,2-pyridinecarboxylic acid, 4-bromo-, methyl ester,4-bromopyridine-2-carboxylic acid methyl ester,abbypharma ap-10-5912,pubchem12733,acmc-209hag,methyl 4-bromo-pyridine-2-carboxylate
12828947
methyl 4-bromopyridine-2-carboxylate
COC(=O)C1=NC=CC(=C1)Br
Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Melting Point 54°C to 56°C
Quantity 5g
Beilstein 473047
Formula Weight 216.03
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material Methyl 4-bromopyridine-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C