Alfa Aesar™ Methyl 4'-chlorobiphenyl-4-carboxylate, 95%

Manufacturer: Alfa Aesar™ H5191006

Quantity 5g

 View more versions of this product

Catalog No. AAH5191006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89901-02-0
    Molecular Formula C14H11ClO2
    Formula Weight 246.69
    MDL Number MFCD04117794
    Synonym 4'-Chlorobiphenyl-4-carboxylic acid methyl ester
    Chemical Name or Material Methyl 4'-chlorobiphenyl-4-carboxylate

    Specifications

    Formula Weight 246.69
    Molecular Formula C14H11ClO2
    MDL Number MFCD04117794
    CAS 89901-02-0
    Synonym 4'-Chlorobiphenyl-4-carboxylic acid methyl ester
    Quantity 5g
    Chemical Name or Material Methyl 4'-chlorobiphenyl-4-carboxylate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit