Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-chlorocarbonylbenzoate, 98+%

Manufacturer:  Alfa Aesar™ H5468806

Quantity 5g

 View more versions of this product

Catalog No. AAH5468806


  Check Availability
Add to cart

Specifications

Description

Applications
4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.

Solubility
Hydrolyzes in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
Description & Specifications

Chemical Identifiers

7377-26-6
198.6
UN3261
Methyl 4-chlorocarbonylbenzoate
C9H7ClO3
MFCD00059486
4-Chlorocarbonylbenzoic acid methyl ester; Methyl 4-(chloroformyl)benzoate

Specifications

7377-26-6
198.6
UN3261
52° to 56°C
Methyl 4-chlorocarbonylbenzoate
C9H7ClO3
MFCD00059486
4-Chlorocarbonylbenzoic acid methyl ester; Methyl 4-(chloroformyl)benzoate
5g
≥98%

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit