Alfa Aesar™ Methyl 4-chlorocarbonylbenzoate, 98+%

Manufacturer: Alfa Aesar™ H5468814

Quantity 25g

 View more versions of this product

Catalog No. AAH5468814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7377-26-6
    Molecular Formula C9H7ClO3
    Formula Weight 198.6
    MDL Number MFCD00059486
    UN Number UN3261
    Synonym 4-Chlorocarbonylbenzoic acid methyl ester; Methyl 4-(chloroformyl)benzoate
    Chemical Name or Material Methyl 4-chlorocarbonylbenzoate

    Specifications

    Formula Weight 198.6
    Molecular Formula C9H7ClO3
    UN Number UN3261
    Synonym 4-Chlorocarbonylbenzoic acid methyl ester; Methyl 4-(chloroformyl)benzoate
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00059486
    CAS 7377-26-6
    Health Hazard 1 Danger
    Melting Point 52° to 56°C
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Methyl 4-chlorocarbonylbenzoate
    Assay ≥98%

    Description

    Applications
    4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.

    Solubility
    Hydrolyzes in water.

    Notes
    Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit