Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-(chloromethyl)benzoate, 97%

Click to view available options
Quantity:
2g
10g
34040-64-7
C9H9ClO2
184.619
MFCD00511224
SATDLKYRVXFXRE-UHFFFAOYSA-N
methyl 4-chloromethyl benzoate,4-chloromethyl benzoic acid methyl ester,methyl-4-chloromethylbenzoate,benzoic acid, 4-chloromethyl-, methyl ester,4-methoxycarbonyl benzyl chloride,methyl alpha-chloro-p-toluate,methyl p-chloromethyl benzoate,p-methoxycarbonyl benzyl chloride,methyl a-chloro-p-toluate,p-toluic acid, .alpha.-chloro-, methyl ester
36664
methyl 4-(chloromethyl)benzoate
COC(=O)C1=CC=C(C=C1)CCl
34040-64-7
C9H9ClO2
184.619
MFCD00511224
SATDLKYRVXFXRE-UHFFFAOYSA-N
methyl 4-chloromethyl benzoate,4-chloromethyl benzoic acid methyl ester,methyl-4-chloromethylbenzoate,benzoic acid, 4-chloromethyl-, methyl ester,4-methoxycarbonyl benzyl chloride,methyl alpha-chloro-p-toluate,methyl p-chloromethyl benzoate,p-methoxycarbonyl benzyl chloride,methyl a-chloro-p-toluate,p-toluic acid, .alpha.-chloro-, methyl ester
36664
methyl 4-(chloromethyl)benzoate
COC(=O)C1=CC=C(C=C1)CCl
Boiling Point 160°C to 165°C (30mmHg)
Quantity 2g
Melting Point 39°C to 40°C
UN Number UN3261
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material Methyl 4-(chloromethyl)benzoate

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.