Alfa Aesar™ Methyl 4-dimethylaminobenzoate, 98+%

Manufacturer: Alfa Aesar™ A1205709

Quantity 10g

 View more versions of this product

Catalog No. AAA1205709

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1202-25-1
    Molecular Formula C10H13NO2
    Formula Weight 179.22
    MDL Number MFCD00014905
    Beilstein 2209703
    Synonym 4-Dimethylaminobenzoic acid methyl ester
    Chemical Name or Material Methyl 4-dimethylaminobenzoate

    Specifications

    Formula Weight 179.22
    Molecular Formula C10H13NO2
    MDL Number MFCD00014905
    CAS 1202-25-1
    Beilstein 2209703
    Melting Point 98° to 102°C
    Quantity 10g
    Synonym 4-Dimethylaminobenzoic acid methyl ester
    Chemical Name or Material Methyl 4-dimethylaminobenzoate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit