Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4'-formylbiphenyl-4-carboxylate, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
70916-89-1
C15H12O3
240.26
URINKTWOCVLGGR-UHFFFAOYSA-N
4'-formylbiphenyl-4-carboxylic acid methyl ester,methyl 4'-formyl-1,1'-biphenyl-4-carboxylate,methyl 4-4-formylphenyl benzoate,methyl 4'-formyl 1,1'-biphenyl-4-carboxylate,acmc-209ogh,methyl 4'-formylbiphenyl-4-carboxylate,methyl-4'-formylbiphenyl-4-carboxylate,1,1'-biphenyl-4,4'-dicarboxylicacid, 4-methyl ester,1,1'-biphenyl-4-carboxylicacid,4'-formyl-,methyl ester
4321323
methyl 4-(4-formylphenyl)benzoate
COC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C=O
70916-89-1
C15H12O3
240.26
URINKTWOCVLGGR-UHFFFAOYSA-N
4'-formylbiphenyl-4-carboxylic acid methyl ester,methyl 4'-formyl-1,1'-biphenyl-4-carboxylate,methyl 4-4-formylphenyl benzoate,methyl 4'-formyl 1,1'-biphenyl-4-carboxylate,acmc-209ogh,methyl 4'-formylbiphenyl-4-carboxylate,methyl-4'-formylbiphenyl-4-carboxylate,1,1'-biphenyl-4,4'-dicarboxylicacid, 4-methyl ester,1,1'-biphenyl-4-carboxylicacid,4'-formyl-,methyl ester
4321323
methyl 4-(4-formylphenyl)benzoate
COC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C=O
Quantity 5g
Melting Point 113.0°C to 118.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 240.26
Percent Purity 97%
Chemical Name or Material Methyl 4'-formylbiphenyl-4-carboxylate

GHS Signal Word: Warning