Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-hydroxybenzoate 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
10kg
Packaging:
Plastic Drum
Plastic bottle
99-76-3
C8H8O3
152.15
MFCD00002352
LXCFILQKKLGQFO-UHFFFAOYSA-N
methylparaben,methyl paraben,methyl p-hydroxybenzoate,nipagin,methyl parahydroxybenzoate,maseptol,p-hydroxybenzoic acid methyl ester,4-hydroxybenzoic acid methyl ester,p-carbomethoxyphenol,tegosept m
7456
CHEBI:31835
methyl 4-hydroxybenzoate
COC(=O)C1=CC=C(C=C1)O
99-76-3
C8H8O3
152.15
MFCD00002352
LXCFILQKKLGQFO-UHFFFAOYSA-N
methylparaben,methyl paraben,methyl p-hydroxybenzoate,nipagin,methyl parahydroxybenzoate,maseptol,p-hydroxybenzoic acid methyl ester,4-hydroxybenzoic acid methyl ester,p-carbomethoxyphenol,tegosept m
7456
CHEBI:31835
methyl 4-hydroxybenzoate
COC(=O)C1=CC=C(C=C1)O
Packaging Plastic bottle
Quantity 500g
Color White
Melting Point 125.0°C to 128.0°C
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Boiling Point 298.6°C
Assay Percent Range 99%
Linear Formula HOC6H4CO2CH3
Beilstein 10,158
Merck Index 15,618
Solubility Information Solubility in water: 0.25% (25°C). Other solubilities: freely soluble in alcohol,acetone and ether,slightly soluble in benzene and ccl4
Formula Weight 152.15
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Methyl 4-hydroxybenzoate, 99%
Show More Show Less

GHS P Statement Harmful to aquatic life with long lasting effects.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-785-7

RTECSNumber : DH2450000

TSCA : TSCA