Alfa Aesar™ Methyl (4-hydroxymethyl)benzoate, 99%

Manufacturer: Alfa Aesar™ B2256414

Quantity 25g

 View more versions of this product

Catalog No. AAB2256414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6908-41-4
    Molecular Formula C9H10O3
    Formula Weight 166.18
    MDL Number MFCD00009984
    Synonym 4-(Hydroxymethyl)benzoic acid methyl ester
    Chemical Name or Material Methyl (4-hydroxymethyl)benzoate

    Specifications

    Formula Weight 166.18
    TSCA TSCA
    Synonym 4-(Hydroxymethyl)benzoic acid methyl ester
    Quantity 25g
    Flash Point >110°C (230°F)
    Molecular Formula C9H10O3
    MDL Number MFCD00009984
    CAS 6908-41-4
    Melting Point 46° to 52°C
    Chemical Name or Material Methyl (4-hydroxymethyl)benzoate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit