Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl (4-hydroxymethyl)benzoate, 99%

Click to view available options
Quantity:
5g
25g
6908-41-4
C9H10O3
166.176
MFCD00009984
VBWFYEFYHJRJER-UHFFFAOYSA-N
methyl 4-hydroxymethyl benzoate,benzoic acid, 4-hydroxymethyl-, methyl ester,4-carbomethoxybenzyl alcohol,4-hydroxymethyl benzoic acid methyl ester,methyl p-hydroxymethyl benzoate,acmc-209o5w,methyl p-hydroxymethylbenzoate,methyl-4-hydroxymethylbenzoate,p-methoxycarbonylbenzyl alcohol
81325
methyl 4-(hydroxymethyl)benzoate
COC(=O)C1=CC=C(C=C1)CO
6908-41-4
C9H10O3
166.176
MFCD00009984
VBWFYEFYHJRJER-UHFFFAOYSA-N
methyl 4-hydroxymethyl benzoate,benzoic acid, 4-hydroxymethyl-, methyl ester,4-carbomethoxybenzyl alcohol,4-hydroxymethyl benzoic acid methyl ester,methyl p-hydroxymethyl benzoate,acmc-209o5w,methyl p-hydroxymethylbenzoate,methyl-4-hydroxymethylbenzoate,p-methoxycarbonylbenzyl alcohol
81325
methyl 4-(hydroxymethyl)benzoate
COC(=O)C1=CC=C(C=C1)CO
Quantity 5g
Flash Point >110°C (230°F)
Melting Point 46°C to 52°C
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Methyl (4-hydroxymethyl)benzoate