Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl 4-iodo-3-nitrobenzoate, 99%
Click to view available options
Quantity:
5g
25g
Description
Applications
Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.
Solubility
Soluble in DCM 10mg/0.5mL. Insoluble in water.
Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.
Solubility
Soluble in DCM 10mg/0.5mL. Insoluble in water.
Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
Specifications
Quantity | 25g |
Sensitivity | Light sensitive |
Melting Point | 101°C to 104°C |
Solubility Information | Soluble in DCM 10mg/0.5mL. Insoluble in water. |
Formula Weight | 307.05 |
Percent Purity | 99% |
Chemical Name or Material | Methyl 4-iodo-3-nitrobenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures