Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-iodosalicylate, 98%

Click to view available options
Quantity:
5g
25g
18179-39-0
C8H7IO3
278.045
MFCD06797864
WUFUURSWOJROKY-UHFFFAOYSA-N
methyl 4-iodosalicylate,2-hydroxy-4-iodobenzoic acid methyl ester,benzoic acid,2-hydroxy-4-iodo-, methyl ester,methyl-4-iodosalicylate,methyl 4-iodosalicyclate,acmc-1c81e,methyl2-hydroxy-4-iodobenzoate,methyl 4-iodo-2-hydroxybenzoate,methyl-2-hydroxy-4-iodo-benzoate
11380407
methyl 2-hydroxy-4-iodobenzoate
COC(=O)C1=C(C=C(C=C1)I)O
18179-39-0
C8H7IO3
278.045
MFCD06797864
WUFUURSWOJROKY-UHFFFAOYSA-N
methyl 4-iodosalicylate,2-hydroxy-4-iodobenzoic acid methyl ester,benzoic acid,2-hydroxy-4-iodo-, methyl ester,methyl-4-iodosalicylate,methyl 4-iodosalicyclate,acmc-1c81e,methyl2-hydroxy-4-iodobenzoate,methyl 4-iodo-2-hydroxybenzoate,methyl-2-hydroxy-4-iodo-benzoate
11380407
methyl 2-hydroxy-4-iodobenzoate
COC(=O)C1=C(C=C(C=C1)I)O
Quantity 25g
Melting Point 68°C to 72°C
Formula Weight 278.05
Percent Purity 98%
Chemical Name or Material Methyl 4-iodosalicylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning